AMMONIUM CHLORIDE 15N 99% NH4CL 54.48 1G
Catalog # MSPP-NLM-467-1
Supplier: Cambridge Isotope Labs Inc
AMMONIUM CHLORIDE 15N 99% NH4CL 54.48 1G
Catalog # MSPP-NLM-467-1
Supplier: Cambridge Isotope Labs Inc
Supplier Number: NLM-467-1
Frequently Bought Together
VWR® Small Latex Bulbs
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Alconox® Powdered Precision Cleaners
Supplier: Alconox
SP Wilmad-LabGlass Thin-Walled Precision NMR Sample Tubes, 5 mm, SP Industries
Supplier: SP Industries
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Trifluoromethanesulfonic anhydride 99%
Supplier: APOLLO SCIENTIFIC
FuGENE® HD Transfection Reagent, Promega
Supplier: Promega Corporation
N-Boc-L-serine
Supplier: APOLLO SCIENTIFIC
Accessories for Q700 Sonicator without a probe, Qsonica
Supplier: QSonica, LLC.