ALKALINE TITRATION SOLVENT 4L GAS/NAPTHA
Catalog # 103790-906
Supplier: Aqua Solutions
ALKALINE TITRATION SOLVENT 4L GAS/NAPTHA
Catalog # 103790-906
Supplier: Aqua Solutions
Supplier Number: 0295-4LG
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Short-Term Detector Tubes, Draeger
Supplier: Draeger
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Hydranal® Coulometric Karl Fischer Titration Reagents, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
STANDARD VISCOSITY GEN PURP S30000 500ML
Supplier: Cannon