Microplate Lids
Microplate lid for MTP/DWP, sterile, Sterile, 80 pcs. (5 bags×16)
Catalog # 89173-635
Supplier: Eppendorf
Supplier #: 0030131525
Jump to:
Product Details & Documents
Protect your samples with Eppendorf® Microplate Lids, perfect for incubation and short-term storage. Stable, flexible protection available in 'PCR Clean' and 'Sterile' grades, ensuring your samples stay pure and uncontaminated.
- Stable, flexible protection of samples during incubation and short-term storage
Specifications
- Sterility:Sterile
- Description:Microplate lid for MTP/DWP, sterile
- Packaging:80 pcs. (5 bags×16)
- Cat. No.:89173-635
- Environmental Certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact