Order Entry
United States
Orders LinkContactUsLinkComponent
Bento Lab

Bento Lab

Bento Lab Basic, 2700 g (fixed), 50 V (fixed)
Catalog # 470323-312
Supplier #:  BLM1B2B
Bento Lab
Catalog # 470323-312
Bento Lab
Bento Lab

Bento Lab Basic, 2700 g (fixed), 50 V (fixed)

Clearance
$1,775.10
Sale

Valid until 31/12/2026

Log in for availability
Each
Product Details & Documents

The mobile lab that includes a thermal cycler, powered electrophoresis, blue light transilluminator, and centrifuge, all into one unit.

  • An entire biotechnology lab the size of a laptop
  • Designed so you can collect samples and check data on the spot
  • No need to connect to a phone, tablet or laptop to edit your protocols
  • Compatible with standard plastic ware and reagents
  • Performance verified by researchers at world-leading research universities

Bento Lab is a portable mobile genomics setup. The compact laboratory is equipped with thermal cycler, microcentrifuge, blue LED transilluminator, gel electrophoresis power supply and electrophoresis gel box. The system is operated using an intuitive on-board graphical interface, and comes with a built-in tube rack.

The thermocycler module: Fast, repeatable PCR results with accurate, uniform block temperature for routine PCR. Equally useful as a block incubator with or without heated lid. Precise, rapid temperature measurements using platinum resistance thermometers, the industry gold standard.

The centrifuge module: Located in the middle of the Bento Lab system, it comes with a see-through, automatically locking lid. The rotor is removable and can accommodate 6×1.5 ml tubes.

The electrophoresis module is made up of two parts. The base is made of clear acrylic, is used to cast gels and as a gel tank during electrophoresis. It also stores casting shutters and combs. The lid is made of orange acrylic that functions as a filter for the blue light transilluminator. The gel box plugs sits on top of the blue LED transilluminator, both of which draw their power directly from the Bento box.

Documents

Specifications
  • Description:
    Bento Lab Basic
  • Electrical:
    50 V (fixed)
  • Details:
    2700 g (fixed)
  • Cat. No.:
    470323-312
  • Gradient:
    -
  • Temperature Range:
    21…99 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.