Order Entry
United States
Orders LinkContactUsLinkComponent
Linearity D-Dimer, AUDIT Microcontrols

Linearity D-Dimer, AUDIT Microcontrols

Linearity FD D-Dimer for bioMerieux VIDAS and miniVIDAS, Freeze dried, D-Dimer, 2 days when stored at 2...8 °C, 5×1 ml
Catalog # 76359-720
Supplier:   Audit Microcontrols
Supplier #:  K845M-5
Linearity D-Dimer, AUDIT Microcontrols
Catalog # 76359-720
CAS Number:  
Linearity D-Dimer, AUDIT Microcontrols
Linearity D-Dimer, AUDIT Microcontrols

Linearity FD D-Dimer for bioMerieux VIDAS and miniVIDAS, Freeze dried, D-Dimer, 2 days when stored at 2...8 °C, 5×1 ml

CAS Number:  
Price
$3,176.22
Log in for availability
Kit of 5 (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Universal linearity D-Dimer, linearity D-Dimer for bioMerieux VIDAS and miniVIDAS, linearity D-Dimer for PATHFAST Immunoanalyzer, linearity D-Dimer for Siemens Stratus are assayed quality control materials consisting of five levels of human plasma.

Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.

Specifications

Specifications

  • Description:
    Linearity FD D-Dimer for bioMerieux VIDAS and miniVIDAS
  • Size:
    5×1 ml
  • Stability:
    2 days when stored at 2...8 °C
  • Analyte:
    D-Dimer
  • Type:
    Freeze dried

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.