ELISA KIT AFP 1.78 IU/ML 96 WELLS 25 UL
Catalog # 76359-628
Supplier: DRG International
ELISA KIT AFP 1.78 IU/ML 96 WELLS 25 UL
Catalog # 76359-628
Supplier: DRG International
Supplier Number: EIA-1468R
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
CellTox Green Cytotoxicity Assay, Promega
Supplier: Promega Corporation