ACETIC ACID/ISOOCTAN 3:2V/V AOCS 4L CS4
Catalog # 103783-138
Supplier: Aqua Solutions
ACETIC ACID/ISOOCTAN 3:2V/V AOCS 4L CS4
Catalog # 103783-138
Supplier: Aqua Solutions
Supplier Number: SPX142
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Glass Fiber Filters
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
Methylene blue 1% (w/v) in aqueous solution
Supplier: Ricca Chemical
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Kerosene, deodorized
Supplier: Aqua Solutions