Electrolyte Solution
Electrolyte 1 M KNO₃, 250 ml, Bottle, pH, ISE, Sulfide, Cadmium, Cyanide, Iodide, Chloride, Thiocyanate, Lead, Copper, Bromide
Supplier: Mettler Toledo
Supplier #: 51350078
Jump to:
Product Details & Documents
pH electrolyte and maintenance solution.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Description:Electrolyte 1 M KNO₃
- Size:250 ml
- For:pH, ISE
- Ion:Sulfide, Cadmium, Cyanide, Iodide, Chloride, Thiocyanate, Lead, Copper, Bromide