WASH BUFFER A SR-X SIMOA 5L BOTTLE PK2
MSPP-103078
Frequently Bought Together
VWR® pH Standards
: VWR
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
DETECTION MODULE ANTI-MOUSE W/PS/SS
: PROTEIN SIMPLE MS
Technical Buffer Solutions, Sachets
: Mettler Toledo
SEPARATION MODULE WES 66-440KDA 8X25 CC
: 0000014126
KIT REPLEX MODULE FOR JESS
: 0000014126
ANTIBODY ANTI-RABBIT SECONDARY
: PROTEIN SIMPLE MS
SGCAA MEDIA WITH GALACTOSE 1000MLSTER
: Teknova
VWR® Vinyl Nitrile Hybrid Examination Gloves
: VWR International
Jess or Wes Separation Module, Protein Simple
: PROTEIN SIMPLE MS
Anti-Rabbit Detection Module, Protein Simple
: PROTEIN SIMPLE MS
WASH BUFFER B SR-X SIMOA 3L BOTTLE PK2
: QUANTERIX CORPORATION MS
EZ STANDARD PACK 1 PK8
: 0000014126
High Sensitivity RNA ScreenTape Analysis
: Agilent Technologies
DILUENT MILK-FREE ANTIBODY 20ML
: 0000014126
EDTA K2 POOLED HU PLASMA BLOOD DERV 50ML
: Innovative Research Inc
PEROXIDE
: 0000014126
ANTIBODY GOAT ANTI-MOUSE HRP CONJUGATE
: PROTEIN SIMPLE MS
Jess or Wes Separation Module, Protein Simple
: 0000014126