DNA/RNA PREP BUFFER CLEAR LIQUID 200ML
Catalog # 76358-838
Supplier: Zymo Research
DNA/RNA PREP BUFFER CLEAR LIQUID 200ML
Catalog # 76358-838
Supplier: Zymo Research
Supplier Number: D7010-2-200
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Supplier: Biotium
Costar® Multiple Well Cell Culture Plates, Corning
Supplier: Corning
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VWR® Powder-Free Nitrile Examination Gloves
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Mitomycin C ≥95%
Supplier: Enzo Life Sciences
96-Well Blocks, Zymo Research
Supplier: Zymo Research