DNA/RNA LYSIS BUFFER 200ML F/QUICK KIT
Catalog # 76358-830
Supplier: Zymo Research
DNA/RNA LYSIS BUFFER 200ML F/QUICK KIT
Catalog # 76358-830
Supplier: Zymo Research
Supplier Number: D7001-1-200
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Axygen® PCR Tubes and Caps, Corning
Supplier: Corning
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International