MAGBINDING BEAD RESEARCH USE ONLY 24ML
Catalog # 76358-798
Supplier: Zymo Research
MAGBINDING BEAD RESEARCH USE ONLY 24ML
Catalog # 76358-798
Supplier: Zymo Research
Supplier Number: D4100-2-24
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Dexamethasone, white powder, Cell Culture Grade
Supplier: MP Biomedicals
RNA Kits
Supplier: Agilent Technologies
RNA Prep Buffer, Zymo Research
Supplier: Zymo Research
Quick-RNA™ Plus Kits, Zymo Research
Supplier: Zymo Research
AMPURE XP REAGENT 60ML
Supplier: 0000038914
MAG BEADDNA/RNA WASH 1 CONCENTRATE 80 ML
Supplier: Zymo Research
Sera-Xtracta™ HMW DNA Kit
Supplier: Cytiva