CELL SCREEN SS 80 MESH 190U 37 MM CS5
Catalog # 76356-930
Supplier: Bellco Glass
CELL SCREEN SS 80 MESH 190U 37 MM CS5
Catalog # 76356-930
Supplier: Bellco Glass
Supplier Number: 1985-00080
Frequently Bought Together
VWR® Standard Standard-Grade Erlenmeyer Flasks
Supplier: VWR International
VWR® Talon® Lab Frame Feet
Supplier: VWR International
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Clear
Supplier: DWK Life Sciences (KIMBLE)
KIMBLE® ULTRA-WARE® 25 mm Microfiltration Assembly with support, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
KONTES® Solid Pennyhead [ST] Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Airfree® Schlenk Stoppers, O-Ring, Chemglass
Supplier: Chemglass
TUBING QUARTZ 7MM ID X 9MM 4FT
Supplier: Chemglass
Glass Burets with Plastic Stopcock
Supplier: United Scientific Supplies
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Supplier: Qorpak
KIMAX® Test Tubes, Plain, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
WHEATON® Serum Vials
Supplier: DWK Life Sciences (KIMBLE)
Accessories for DURAN® Bottle GLS® 80 Connection System, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Pluro Stopper, Neoprene, Ace Glass Incorporated
Supplier: Ace Glass
Media Bottles, Graduated, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Cell Culture Flask, GL 45, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Black Phenolic Screw Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
DURAN® YOUTILITY, Bottle Tags, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Graduated Cylinders, Glass, Class A, Individually Certified, with Stopper, United Scientific Supplies
Supplier: United Scientific Supplies
CELL SCREEN SS 20 MESH 860U 37 MM CS5
Supplier: Bellco Glass