Order Entry
United States
ContactUsLinkComponent
CELL SCREEN SS 50 MESH 280U 37 MM CS5
Catalog # 76356-926
Supplier:  Bellco Glass
undefined
CELL SCREEN SS 50 MESH 280U 37 MM CS5
Catalog # 76356-926
Supplier:  Bellco Glass
Supplier Number:  1985-00050
Frequently Bought Together