Order Entry
United States
ContactUsLinkComponent
IOB W/MICRO-CARRIER FLASK 100MM 45MM 8L
Catalog # 76356-628
Supplier:  Bellco Glass
undefined
IOB W/MICRO-CARRIER FLASK 100MM 45MM 8L
Catalog # 76356-628
Supplier:  Bellco Glass
Supplier Number:  1965-61080
Frequently Bought Together