COVERSLIP ROUND GERMAN GLASS 12MM CS100
Catalog # 76355-906
Supplier: Bellco Glass
COVERSLIP ROUND GERMAN GLASS 12MM CS100
Catalog # 76355-906
Supplier: Bellco Glass
Supplier Number: 1943-10012A
Frequently Bought Together
PBS (Phosphate Buffered Saline), 10X, pH 7.4, GrowCells
Supplier: GROWCELLS
VWR® Caps
Supplier: VWR International
VWR® Aluminum Micro Weighing Dishes
Supplier: VWR International
Round Cover Slips, German Glass, Bellco Glass
Supplier: Bellco Glass
Calpain Inhibitor 1
Supplier: Thermo Scientific Chemicals
Guanidinium hydrochloride ≥98%
Supplier: BeanTown Chemical
Substance P ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) 98+%
Supplier: Thermo Scientific Chemicals
COVERSLIP ROUND GERMAN GLASS 15 MM CS100
Supplier: Bellco Glass
VWR® Pressure Sensitive Color-Coded Labeling Tapes
Supplier: VWR International
(3-Mercaptopropyl)trimethoxysilane ≥95%
Supplier: BeanTown Chemical
Wheat Germ Agglutinin (WGA) Conjugates, Biotium
Supplier: Biotium
German Glass Coverslips, PDL Coated, Neuvitro Corporation
Supplier: NEUVITRO CORPORATION MS
1 kb Plus DNA Ladder for Safe Stains, New England Biolabs
Supplier: New England Biolabs (NEB)
Toluene, anhydrous ≥99.5%
Supplier: BeanTown Chemical
REVERT™ Total Protein Stain Kits, LI-COR®
Supplier: Li-Cor
Magne Protein G Beads, 20% Slurry, Promega
Supplier: Promega Corporation