Order Entry
United States
Orders LinkContactUsLinkComponent
Nitrofurantoin

Nitrofurantoin

1 g
Catalog # 76347-320
Irritant
Health
Supplier:   RPI
Supplier #:  N51500-1.0
Nitrofurantoin
Catalog # 76347-320
Supplier:  RPI
CAS Number:  67-20-9
Nitrofurantoin
Nitrofurantoin

1 g

CAS Number:  67-20-9
Price
$35.78
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.

Researchers may use nitrofurantoin as a selective agent by incorporating it into culture media to promote the growth of cells or organisms that have been genetically engineered to express resistance to this antibiotic. It may have applications in microbiology and cell culture experiments, particularly in the context of antibiotic selection.

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  1-[(5-Nitrofurfurylidene)amino]hydantoin
  • CAS Number:67-20-9
  • Formula:C₈H₆N₄O₅
  • MDL Number:MFCD00003224
  • Molecular Weight:238.16 g/mol
  • Melting Pt:263...265 °C
  • Storage Temperature:Room Temperature
  • EINECS:200-646-5
Specifications

Specification Test Results

  • Appearance:
    Yellow crystalline powder
  • Assay:
    98.0 - 102.0 %
  • Identification:
    Pass test
  • Sulfated Ash:
    ≤ 0.1 %
  • Related Substances:
    ≤ 1.0 %

Specifications

  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.