Order Entry
United States
Orders LinkContactUsLinkComponent
Nalidixic acid

Nalidixic acid

1 kg
Catalog # 76347-288
Health
Supplier:   RPI
Supplier #:  N42000-1000.0
Nalidixic acid
Catalog # 76347-288
Supplier:  RPI
CAS Number:  389-08-2
Nalidixic acid
Nalidixic acid

1 kg

CAS Number:  389-08-2
Price
$1,347.84
Log in for availability
Each (1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Nalidixic Acid Sodium Salt is a chemical compound that is used for its antimicrobial properties and has various applications in microbiology, and pharmaceutical research. Nalidixic Acid Sodium Salt is employed as a reference standard and selective agent in microbiological culture media. It inhibits the growth of susceptible bacteria, making it a valuable tool for studying bacterial susceptibility and resistance patterns.

Researchers use it to assess the effectiveness of antibiotics against different bacterial strains, contributing to our understanding of microbial biology and antibiotic resistance.

In pharmaceutical research, Nalidixic Acid Sodium Salt can be used as a reference standard for antibiotic testing and formulation. It supports the development and optimization of antibiotics and strategies to combat antibiotic-resistant bacteria.

Caution: Research or further manufacturing use only, not for food or drug use.

  • CAS Number:389-08-2
  • Formula:C₁₂H₁₂N₂O₃
  • MDL Number:MFCD00006884
  • Molecular Weight:232.24 g/mol
  • Melting Pt:227…229 °C
  • Melting Pt:227 - 229 °C
  • Storage Temperature:Room Temperature
  • EINECS:206-864-7
Specifications

Specification Test Results

  • Appearance:
    White to very pale yellow, odorless crystalline powder
  • Solubility:
    Soluble in chloroform, in methylene chloride, and in solution of fixed alkali hydroxides and carbonates; slightly soluble in acetone, in alcohol, in methanol and in toluene; very slightly soluble in ether and in water.
  • Identification (Infrared Absorption):
    Complies
  • Identification (Ultraviolet Absorption) @258nm on dry basis:
    ≤ 3.0
  • Melting Range:
    225 - 231 oC
  • Loss on Drying:
    ≤ 0.5 %
  • Residue on Ignition:
    ≤ 0.1 %
  • Chromatographic Purity (By TLC):
    Complies
  • Assay (Dry Basis):
    99.0 - 101.0 %
  • BSE/TSE Free:
    Conforms
  • Grade:
    The material complies as per USP specification.

Specifications

  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.