Order Entry
United States
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol

Plastic bottle, 1 kg
Catalog # 76346-838
Supplier:   RPI
 :  M22080-1000.0
D-(-)-Mannitol
Catalog # 76346-838
Supplier:  RPI
CAS Number:  69-65-8
D-(-)-Mannitol
D-(-)-Mannitol

Plastic bottle, 1 kg

CAS Number:  69-65-8
Price
$73.58
Log in for availability
Each (1kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : Research or further manufacturing use only, not for food or drug use.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Melting Pt:167...170 °C
  • Storage Temperature:Room Temperature
  • EINECS:200-711-8
Specifications

Specification Test Results

  • Appearance:
    White crystalline powder
  • Appearance of Solution:
    Clear and colourless solution
  • Melting Point:
    165.0 - 170.0 °C
  • Nickel (Ni):
    ≤ 1 ppm
  • Conductivity:
    ≤ 20 µS/cm
  • Total Yeast and Mold Count (TYMC):
    ≤ 100 cfu/g
  • Identification (IR):
    Conforms
  • Reducing Sugar:
    ≤ 0.1 %
  • Loss on Drying:
    ≤ 0.5 %
  • Residual Solvents:
    Meet requirements
  • Bacterial Endotoxins:
    ≤ 2.5 IU/g
  • E.Coli:
    Negative
  • Total Aerobic Microbial Count (TAMC):
    ≤ 100 cfu/g
  • Assay:
    96.0 - 101.5 %
  • Related Substance ( Maltitol and Isomalt):
    ≤ 2.0 %
  • Related Substance (Sorbitol):
    ≤ 2.0 %
  • Related Substance (Total impurity):
    ≤ 2.0 %
  • Related Substance ( Unspecified Impurities):
    ≤ 0.10 %
  • Specific Optical Rotation (EP):
    +23 - +25 °
  • Specific Optical Rotation (USP):
    +137 - +145 °
  • Non Animal Origin:
    Conforms
  • Grade:
    USP/EP

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.