D-(+)-Mannose
Plastic bottle, 500 g
Catalog # 76346-758
Supplier: RPI
Supplier #: M18000-500.0
Jump to:
Product Details & Documents
Caution: Research or further manufacturing use only, not for food or drug use.
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Storage Temperature:Room Temperature
- EINECS:222-392-4
Specifications
Specification Test Results
- Appearance:White powder/off white powder
- Assay:≥99.0%
- Heavy Metals:≤10 ppm
- Loss on Drying:≤0.5%
- Melting Point:124 - 136 °C
- Optical Rotation:+13.0 - +15.0°
- Yeast and Mold:≤1000 cfu/g
- BSE/TSE Free:BSE/TSE Free
- Country of Origin:Report
Specifications
- Pack type:Plastic bottle
- Size:500 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



