CARTRIDGE CAPILLARY PRE-ASSEMBLED
MSPP-A55625
Frequently Bought Together
Triethylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Disposable Microcentrifuge Tubes
: VWR International
ASSAY KIT IGG PURITY/HETEROGENEITY
: 0000013719
APERTURE CAPILLARY CARTRID 100X800UM PK3
: 0000013719
Accessories for Densito and DensitoPro Handheld Density Meters
: Mettler Toledo
Accessories for METTLER TOLEDO Titrators
: Mettler Toledo
BUFFER VIAL TRAY 6X6
: 0000013719
SCREWCAP WITH SEPTUM FOR 8ML GLASS PK100
: METROHM USA MS
Zenix™ SEC Columns, Sepax Technologies
: Sepax Technologies
Universal Vial Caps, Ab Sciex
: 0000013719
Sodium hydroxide 10.0 N , from NF/USP material
: Ricca Chemical
INTERNAL STANDARD 10 KD MARKER
: 0000013719
CARTRIDGE ASSEMBLY 30 CM CAPILLARY
: 0000013719
Universal Vials, Ab Sciex
: 0000013719
BUFFER SAMPLE LOW PH SDS 140ML
: 0000013719
CABLE FIBER OPTIC FOR PDA DETECTOR
: 0000013719
UniStore™ Sterile Media Bottles, PETG
: United Scientific Supplies