KIT QUICK WESTERN IRDYE 680RD
Catalog # 103766-354
Supplier: Li-Cor
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® TBS Buffer, 20X Liquid
Supplier: VWR
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Acrylamide and Bis-Acrylamide
Supplier: VWR
VWR® Universal Pipette Tips
Supplier: VWR International
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
60% SUCROSE SOLUTION1000MLENDOTOXIN
Supplier: Teknova
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
Flow Cell Wash Kits
Supplier: Oxford Nanopore Technologies
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza