T20 PBS ANTIBODY DILUNT W/TWEEN20 1500ML
Catalog # 103761-218
Supplier: Li-Cor
T20 PBS ANTIBODY DILUNT W/TWEEN20 1500ML
Catalog # 103761-218
Supplier: Li-Cor
Supplier Number: 927-75003
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
RIPA Lysis Buffer, Rockland
Supplier: Rockland Immunochemical
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
BUFFER PBS BLOCKING READY-TO-USE 1500ML
Supplier: Li-Cor