T20 TBS ANTIBODY DILUNT W/TWEEN20 5000ML
Catalog # 103761-212
Supplier: Li-Cor
T20 TBS ANTIBODY DILUNT W/TWEEN20 5000ML
Catalog # 103761-212
Supplier: Li-Cor
Supplier Number: 927-65010
Frequently Bought Together
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Ethanol ≥70% (v/v), Biotechnology Grade
Supplier: VWR
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
NEBUILDER HiFi DNA Assembly® Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)