Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER (PBS) RECONSTITUTION 2 ML

BUFFER (PBS) RECONSTITUTION 2 ML

Catalog # 103745-704
Marketsource
Supplier:   R&D Systems
Supplier #:  RB01
BUFFER (PBS) RECONSTITUTION 2 ML
Catalog # 103745-704
Supplier:  R&D Systems
Marketsource
CAS Number:  
BUFFER (PBS) RECONSTITUTION 2 ML
CAS Number:  
Price
Inquire For Price
Log in for availability
Each (2ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents