Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER TBS BLOCKING READY-TO-USE 5000ML

BUFFER TBS BLOCKING READY-TO-USE 5000ML

Catalog # 103749-020
Marketsource
Supplier:   Li-Cor
Supplier #:  927-60010
BUFFER TBS BLOCKING READY-TO-USE 5000ML
Catalog # 103749-020
Supplier:  Li-Cor
Marketsource
CAS Number:  
BUFFER TBS BLOCKING READY-TO-USE 5000ML
CAS Number:  
Price
$1,372.31
Log in for availability
Each(500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents