Orders LinkContactUsLinkComponent
Mini Block Heaters
Mini Block Heaters
Block, fits 24 tubes, 0.5 ml
Catalog # 470319-524
Supplier #:  18900415
Mini Block Heaters
Catalog # 470319-524
Mini Block Heaters
Mini Block Heaters
Block, fits 24 tubes, 0.5 ml
Price
$158.58
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Affordable mini dry-block heaters are a great option for the lab.
  • LCD Display of both temperature and time
  • Overheating protection
  • Rapid, 3-point self-calibration
  • USB Interface to save data
  • Available with or without lid/cooling
These single block mini heaters come with a block that can hold fifteen, 1.5 ml tubes, but with various additional blocks available, most common tubes can be used in these units. Three different varieties allow to choose whether or not you want to utilize a heated lid, and whether you want heating or heating and cooling. Choose the machine that best suits your needs.
Specifications
  • Description:
    Block, fits 24 tubes
  • For tube volume:
    0.5 ml