Phenol Red Dextrose Broth
500 g
Catalog # 470317-440
Supplier: Ward's Science
Supplier #: AD-MM1038-500G
Jump to:
Product Details & Documents
To prepare media, add 20 g of phenol red dextrose broth to a liter of distilled or de-ionized water. The media will not be sterile until autoclaved prior to use.
Specifications
Specifications
- Size:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



