FRIT PE 10UM 1/16 12G FLASH COLUMNPK20
Catalog # 76328-830
Supplier: Phenomenex
FRIT PE 10UM 1/16 12G FLASH COLUMNPK20
Catalog # 76328-830
Supplier: Phenomenex
Supplier Number: FCS012-SX
Frequently Bought Together
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Closures for Headspace Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
GC Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
SafetyCaps
Supplier: SCAT AMERICAS, INC.
Vaplock™ Evaporation Control and Relief Valves, Cole-Parmer
Supplier: Antyila Scientific
Extraction Kits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Vaplock™ Justrite Container Quick Disconnect Waste Caps and Waste Kits, Cole-Parmer
Supplier: Antyila Scientific
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
PTFE Tubing, PerkinElmer
Supplier: PerkinElmer
Accessories for SCAT SafetyCaps, SCAT Europe
Supplier: SCAT AMERICAS, INC.
Pump Valves and Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Closures for 2 ml Crimp/Snap Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Elizabethkingia meningoseptica Recombinant N-Glycanase (PNGase F) (from E. coli)
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Metering Device Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Agilent Autosampler Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Superloops, 10 and 50 ml, Cytiva
Supplier: Cytiva