Basic IV Administration Sets, B. Braun
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Documents
Specifications
- Flow rate:15 Drops/ml
- Sterility:Sterile
- Description:2 Universal spikes, 2 roller clamps, injection site, SPIN-LOCK® Connector, Priming Volume: 25 ml
- Length:218 cm (86")