Orders LinkContactUsLinkComponent
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Rate Flow Regulator with Female Luer Connector, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 2.6 ml, 46 cm (18")
Catalog # 76327-166
Supplier #:  V5200
Basic IV Administration Sets, B. Braun
Catalog # 76327-166
Supplier:  
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Rate Flow Regulator with Female Luer Connector, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 2.6 ml, 46 cm (18")
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Specifications
  • Description:
    Rate Flow Regulator with Female Luer Connector, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 2.6 ml
  • Length:
    46 cm (18")