Baseline® Windmill Spirometers
Paper Mouthpieces
Catalog # 470316-652
Supplier: Fabrication Enterprises
Supplier #: 12-1714
Jump to:
Product Details & Documents
Compact and lightweight.
- Replacement mouthpieces in plastic or paper
Compact, lightweight spirometer for screening large populations. Subject blows into disposable mouthpiece. Indicator registers forced vital lung capacity up to 7 liters.
Delivery information: Includes 50 mouthpieces.
Documents
Specifications
- Description:Paper Mouthpieces