Orders LinkContactUsLinkComponent
Baseline® Windmill Spirometers
Baseline® Windmill Spirometers
Windmill Spirometer
Catalog # 470316-648
Supplier #:  12-1710
Baseline® Windmill Spirometers
Catalog # 470316-648
Baseline® Windmill Spirometers
Baseline® Windmill Spirometers
Windmill Spirometer
Price
$225.30
Log in for availability
Each Case of 10
Product Details & Documents
Compact and lightweight.
  • Replacement mouthpieces in plastic or paper
Compact, lightweight spirometer for screening large populations. Subject blows into disposable mouthpiece. Indicator registers forced vital lung capacity up to 7 liters.
Delivery information: Includes 50 mouthpieces.
Specifications
  • Description:
    Windmill Spirometer