Orders LinkContactUsLinkComponent
Ward's® Essentials Research Binocular Microscopes
Ward's® Essentials Research Binocular Microscopes
Compound Binocular Microscope, LED
Catalog # 470315-508
Supplier:   VWR
Supplier #:  SW380B
Ward's® Essentials Research Binocular Microscopes
Catalog # 470315-508
Supplier:  VWR
Ward's® Essentials Research Binocular Microscopes
Ward's® Essentials Research Binocular Microscopes
Compound Binocular Microscope, LED
Price
$354.15
Log in for availability
Each
Product Details & Documents
Packed with standard research features, these microscopes are ideal for educational and professional applications.
  • Professional Siedentoph binocular head
  • Six magnification levels: 40×, 100×, 250×, 400×, 1000× and 2500×
  • 10× and 25× interchangeable wide-field eyepieces
  • 1 year limited warranty
Combining an ergonomic design and advanced features this microscope fits the needs of student and clinical environments. Ultra-precise focusing and built-in mechanical stage contribute to the ease of use and the glass optics and cool adjustable LED illumination provide clear, crisp images.
The 1.3MP USB camera includes user-friendly editing and processing software compatible with Windows XP, Vista, 7, 8 and 10.
Specifications
  • Description:
    Compound Binocular Microscope
  • Illumination:
    LED