MEDIA ASTROCYTE CONDITIONED 100ML
Catalog # MSPP-1811
Supplier: SCIENCELL RESEARCH LAB MS
MEDIA ASTROCYTE CONDITIONED 100ML
Catalog # MSPP-1811
Supplier: SCIENCELL RESEARCH LAB MS
Supplier Number: 1811
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
PIG® Universal Pads and Rolls, New Pig
Supplier: New Pig
VWR® Disposable Petri Dishes
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® Charcoal-Stripped FBS, Corning
Supplier: Corning
PROTEIN HUMAN LAMININ 521 LA5H52611MG
Supplier: ACROBIOSYSTEMS