ANTIBODY CD3 LN10 BOND RTU PRIMARY 30ML
Catalog # MSPP-PA0122
Supplier: Leica Biosystems MS
ANTIBODY CD3 LN10 BOND RTU PRIMARY 30ML
Catalog # MSPP-PA0122
Supplier: Leica Biosystems MS
Supplier Number: PA0122
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® Razor Blades
Supplier: VWR International
Acetone ≥99.5%, Epredia™ for histology Dehydrant
Supplier: Epredia
Cell Culture Storage Bottles, Polystyrene, Corning
Supplier: Corning
VWR® Sterile 70% Isopropanol
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Anti-CEACAM6 Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
MAB CHO CELLS PEMBROLIZUMAB HU 98% 5MG
Supplier: Selleck Chemicals
KIT ASPIRATING PRB CLNG BOND F/15 CLEANS
Supplier: Leica Biosystems MS