ANTIBODY CD163 BOND RTU PRIMARY 7ML
Catalog # MSPP-PA0090
Supplier: Leica Biosystems MS
ANTIBODY CD163 BOND RTU PRIMARY 7ML
Catalog # MSPP-PA0090
Supplier: Leica Biosystems MS
Supplier Number: PA0090
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Disposable Transfer Pipettes
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
DILUENT BONDTM PRIMARY ANTIBODY 0.5L
Supplier: Leica Biosystems MS