VWR® Traceable® Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Product Details & Documents
Easily access collected data between destinations with Bluetooth® data loggers.
- Connect to data via smartphone or tablet (any Bluetooth-enabled device) using a downloadable Bluetooth App
- View and download data with free TraceableGO™ App
- Data loggers store up to 90 alarm events
- Readings and setup parameters easily viewable on large 2×1" LCD
- Get accurate, reliable measurements with included NIST-traceable calibration
- One-year warranty
- Compatible with TraceableGO or TraceableLIVE mobile apps
- Critical Information(Temperature, trip duration, MIN/MAX, battery level and alarms) all displayed on LCD at all times
Users have the option to subscribe to cloud-based TraceableLIVE® service to upload data to cloud, which is ideal for tracking data from multiple loggers or to comply with regulatory agency requirements. The low-cost TraceableLIVE® subscription provides unlimited trip data to be saved on the cloud as well as the ability to send a report to an email.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as they are received. Each product goes through an intense vetting process before it can carry the Traceable® name. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving both time and money by not having to calibrate separately.
Bluetooth device is the ideal transit partner for monitoring materials between destinations. Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location. TraceableGO mobile app immediately identifies if the contents experienced conditions outside user-defined parameters. Broad compatibility easy bluetooth connection works with any bluetooth-enabled devices.
With the TraceableGO mobile app, view if an alarm was triggered, generate an instant data report. Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours, logging duration is 227 days (7.5 months). TraceableGO™ Bluetooth devices are pre-programmed with alarm values.
Certifications: Made in the USA; CE marked; NIST-traceable calibration also helps users meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA, and Joint commission requirements. 21 CFR Part 11 compliance with TraceableLIVE App. Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from an ISO/IEC 17025:2017 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement.
Delivery information: Model with bullet probe includes bullet probe, two AAA batteries, and NIST-traceable calibration certificate with data. Model with bottle probe includes bottle probe, two AAA batteries, and NIST-traceable calibration certificate with data. Model with RTD probe includes 10' (3 m), PFA coated with stainless steel tip, RTD probe, two AAA batteries, and NIST-traceable calibration certificate with data.
Documents
VWR TraceableGO Data Logger Thermometers with External Probes
Product Literature
Specifications
- Resolution:T: 0.1 °C
- Description:Temperature data logger with bullet probe
- Accuracy:T: ±0.3 °C (±0.5 °F)
- Range:T: −50...+70 °C (−58…+158 °F)
- Cat. No.:76321-888
- Data Memory:65536
- Recording intervals:1 min - 12 h
- W×D×H:89×20×51 mm (3.5×0.79×2")
- Batteries:AAA Batteries
- Interface:Bluetooth
- Weight:0.65 oz.
- Display:Multiline LCD



