Orders LinkContactUsLinkComponent
Camera Adapters
Camera Adapters
Photo adapter, 2.5× and 4.0× eyepiece, —
Catalog # 470314-908
Supplier:   Motic
Supplier #:  1101001901931
Camera Adapters
Catalog # 470314-908
Supplier:  Motic
Camera Adapters
Photo adapter, 2.5× and 4.0× eyepiece, —
Price
$160.40
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Specifications
  • Description:
    Photo adapter, 2.5× and 4.0× eyepiece
  • For use with:
Recommendations