Orders LinkContactUsLinkComponent
Camera Adapters
Camera Adapters
C-mount adapter for 2/3" chip sensors, 0.65×, Connects with AE31E/AE2000, BA series and SMZ161/171 series
Catalog # 470314-904
Supplier:   Motic
Supplier #:  1101001901781
Camera Adapters
Catalog # 470314-904
Supplier:  Motic
Camera Adapters
C-mount adapter for 2/3" chip sensors, 0.65×, Connects with AE31E/AE2000, BA series and SMZ161/171 series
Price
$199.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Specifications
  • Description:
    C-mount adapter for 2/3" chip sensors, 0.65×
  • For use with:
    Connects with AE31E/AE2000, BA series and SMZ161/171 series