Ward's® Pure Preserved Squids
Product 470314-746 is a replacement for 470314-632 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Ward's® PURE Preserved Specimens - the best dissection experience from start to finish.
- Safest specimens
- Low formalin odor
- Best classroom experience
Ward's® PURE preserved specimens offer the best classroom experience because PURE specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Loligo or ommastrephes. The squid has the most highly developed nervous system of all the invertebrates. Dissection shows developed eyes and nervous system.
Specifications
- Description:Pure preserved squid
- Size:12"+
- Injection type:Double injected
- Packaging:Pail of 10