Order Entry
United States
Orders LinkContactUsLinkComponent
VWR® E-Burette

VWR® E-Burette

VWR® E-Burette with motor piston, 50 ml, ±0.05%, ±0.02%, 10 µl
Catalog # 76319-574
Supplier:   VWR International
VWR® E-Burette
Catalog # 76319-574
Supplier:  VWR International
VWR® E-Burette
VWR® E-Burette

VWR® E-Burette with motor piston, 50 ml, ±0.05%, ±0.02%, 10 µl

Price
$1,735.07
New Customer Promotional Price
Log in for availability
Each (50ml)
Product Details & Documents

State-of-the art motor operated burette with three calibrated pre-set speeds and accuracy and precision conforming to ISO 8655 standards.

  • Motor controlled piston movement
  • Can be flexibly operated at 3 calibrated pre-set speeds: Fast, medium and dropwise
  • Recirculation mode to prevent the loss of reagent during purging
  • Flexible delivery nozzle to change angle of dispensing
  • User-friendly TFT touchscreen
  • Hand held stylus provided as an accessory to assist touch operations
  • Powered by rechargeable, replaceable long lasting Li-ion battery
  • Up to 20 results can be stored in the instrument memory
  • Computer connectivity through a micro USB cable

The VWR® E-Burette is a motor operated burette with sophisticated features and functionality. Motor controlled piston movement eliminates the errors associated with manual wheel controlled piston movement, thus facilitating correct and comfortable titrations. Having the capacity of 50 ml, this burette can be flexibly operated at 3 calibrated pre-set speeds, including dropwise dispensing for fine volume adjustment. This helps in achieving accurate end point of the titration experiment.

The VWR® E-Burette has a ‘recirculation mode’ which prevents loss of reagents during purging and facilitates bubble-free dispensing. The delivery nozzle of the burette can be adjusted in both horizontal and vertical directions providing easy dispensing in demanding laboratory conditions.

The user-friendly TFT touchscreen guides quick and accurate titrations and displays numeric data up to the second decimal. A hand held stylus is provided as an accessory to assist touch operations on the control panel. The touch screen control panel can be used to perform functions like automatic re-fill without change of reading, zero reset, fill and dispense. The control panel is powered by a rechargeable, long lasting Li-ion battery. This battery can be easily replaced by the user.

The instrument has computer connectivity through a mini USB cable. It has a built-in software which allows readings to be transferred and saved in a file on the computer.

Certifications: Meets ISO 8655 standard for accuracy and precision, manufacturing standards: ISO 9001, ISO 13485, ISO 17025 and product certification: CE

Ordering information: Supplied with a digital bottle top burette, a control panel, a micro-USB cable, telscoping tube, five thread adapters (28, 32, 38, 40 and 45 mm), stylus, amber colored window, user instruction manual and an individual quality control certificate that documents the serial number and test values, and confirms the conformity according to weights and measures regulations as per ISO 8655.

Caution: Maintain instrument and reagent temperature between +10 to +40 °C (From 50 to 104 °F). Maintain reagent vapor pressure up to 600 mbar (maximum). Aspirate slowly above 300 mbar pressure in order to prevent the liquid from boiling. Kinematic viscosity up to 500 mm²/s. Relative air humidity between 20 and 90%. Use fluids with density up to 2.2 g/cm³.

Specifications
  • Volume:
    50 ml
  • Description:
    VWR® E-Burette with motor piston
  • Accuracy:
    ±0.05%
  • Precision:
    ±0.02%
  • Subdivisions:
    10 µl
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • VWR® Bottle top dispenser adapter assortment, sizes 25, 28, 32, 45 mm
    VWR® Bottle top dispenser adapter assortment, sizes 25, 28, 32, 45 mm
    76319-594
    Log in for availability
    Pack of 4 (1/Itm/Bx) - $42.64

Product Information

  • Catalog #
    Availability
    Price
  • VWR® Bottle top dispenser adapter, size 32 mm
    VWR® Bottle top dispenser adapter, size 32 mm
    76319-596
    Log in for availability
    Pack of 3 (1/Itm/Bx) - $44.26

Product Information

  • Catalog #
    Availability
    Price
  • VWR® Bottle top dispenser adapter, size 45 mm
    VWR® Bottle top dispenser adapter, size 45 mm
    76319-598
    Log in for availability
    Pack of 3 (1/Itm/Bx) - $36.17

Product Information

  • Catalog #
    Availability
    Price
  • VWR® Bottle top dispenser adapter, size 38 mm
    VWR® Bottle top dispenser adapter, size 38 mm
    76319-602
    Log in for availability
    Pack of 3 (1/Itm/Bx) - $37.91

Product Information

  • Catalog #
    Availability
    Price
  • VWR® Bottle top dispenser adapter, size 40 mm
    VWR® Bottle top dispenser adapter, size 40 mm
    76319-600
    Log in for availability
    Pack of 3 (1/Itm/Bx) - $36.17

Product Information

  • Catalog #
    Availability
    Price
  • VWR® Bottle top dispenser adapter, size 25 mm
    VWR® Bottle top dispenser adapter, size 25 mm
    76319-606
    Log in for availability
    Pack of 3 (1/Itm/Bx) - $36.17

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.