Order Entry
United States
Orders LinkContactUsLinkComponent
Ward's Pipette Racks

Ward's Pipette Racks

Ward's ABS pipette rack, 291×88×401 mm, Clear
Catalog # 470311-172
Supplier:   Heathrow Scientific
Supplier #:  HS20611W
Ward's Pipette Racks
Catalog # 470311-172
Ward's Pipette Racks
Ward's Pipette Racks

Ward's ABS pipette rack, 291×88×401 mm, Clear

Price
$130.45
Log in for availability
Each
Product Details & Documents

Protects and organizes glass and plastic pipettes.

  • Cut out viewing window
  • Available in magnetized option

Angled four shelved compartments keep 0.1 to 50 ml pipettes or cans within easy reach while preventing damage and clutter. Each pipette rack comes with two removeable attachments that hold pipette bulbs, fillers, and pumps. The Magnetized version has strong magnets on both sizes and mounts securely to most metal surfaces.

Specifications
  • Color:
    Clear
  • Description:
    Ward's ABS pipette rack
  • Dimensions:
    291×88×401 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.