SUPPLEMENT TRANSFECTORY BROWN 500ML
Catalog # MSPP-91148500
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
SUPPLEMENT TRANSFECTORY BROWN 500ML
Catalog # MSPP-91148500
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Supplier Number: 91148-500ML
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
AcroPak™ Filtration Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Cell Culture Storage Bottles, Polystyrene, Corning
Supplier: Corning
VWR® Sterile 70% Isopropanol
Supplier: VWR International
SP Bel-Art Poxygrid® Bench-Top Biohazard Bag Holder Kit, Bel-Art Products, a Part of SP
Supplier: Bel-Art Products, a Part of SP
VWR® Disposable Petri Dishes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
SUPPLEMENT ANTI-CLUMPING LIQUID 50ML
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
L-GLUTAMINE LIQUID 200MM 100X 100ML
Supplier: Invitrogen
FectoCHO® Expression System
Supplier: Polyplus Transfection
SATURATED AMMONIUM SULFATE SOLUTION
Supplier: Teknova
BALANCD TRANSFECTORY CHO LIQUID 1L
Supplier: FUJIFILM IRVINE SCIENTIFIC MS