BILIRUBIN ASSAY KIT COLORIMETRIC 200 ASY
Catalog # 103721-694
Supplier: Cell Biolabs
BILIRUBIN ASSAY KIT COLORIMETRIC 200 ASY
Catalog # 103721-694
Supplier: Cell Biolabs
Supplier Number: MET-5010
Frequently Bought Together
VWR® Dissecting Scissors, Sharp Tip, 4¹/₂"
Supplier: VWR International
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Corning® Centrifuge Tubes, 15 ml
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: Hardy Diagnostics
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek
Lok-Tight™ Contact Plates for Surface Sampling, Hardy Diagnostics
Supplier: Hardy Diagnostics
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Supplier: Hardy Diagnostics
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies