REAGENT CYTOLIGHT RAPID RED 1MM 20UL PK5
Catalog # MSPP-4706
Supplier: SARTORIUS CORPORATION MS
REAGENT CYTOLIGHT RAPID RED 1MM 20UL PK5
Catalog # MSPP-4706
Supplier: SARTORIUS CORPORATION MS
Supplier Number: 4706
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Films for ELISA and General Incubation, Polyester
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Bovine Thrombin (from Plasma), MP Biomedicals
Supplier: MP Biomedicals
ASSAY UPLEX HUMAN IL17A K151UTK-2
Supplier: Meso Scale Diagnostics