Order Entry
United States
Orders LinkContactUsLinkComponent
 

Potassium hydroxide solution 2N

1 L
Catalog # 76287-212
Supplier:   G-Biosciences
 :  786-819
Potassium hydroxide solution 2N
Catalog # 76287-212
Supplier:  G-Biosciences
CAS Number:  1310-58-3
Potassium hydroxide solution 2N

1 L

CAS Number:  1310-58-3
Price
$597.34
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A strong, standardized alkaline solution commonly used in biochemistry for pH adjustment, tissue digestion, protein hydrolysis, and cell lysis.

  • Synonym: Caustic potash CAS number: 1310-58-3 Molecular formula: KOH Molecular weight: 56.11

Its strong base properties make it essential in various analytical and preparative procedures where precise control of alkalinity is required.

Synonyms:  Potash caustic, Potassium hydrate, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
Specifications

Specifications

  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.