Orders LinkContactUsLinkComponent
Gentamicin sulfate USP
Bottle, 10 g
  76285-826
 :  RC-173
Gentamicin sulfate USP
  76285-826
 :  1405-41-0
Gentamicin sulfate USP
Bottle, 10 g
 :  1405-41-0
$147.25
Each
Gentamicin efficiently inhibits growth of many Gram-positive and Gram-negative microorganisms. It does so by inhibiting protein synthesis by binding to the L6 protein of the 30S ribosomal subunit. It is also effective against mycoplasma. Recommended for use in cell culture applications.
  • USP Grade
 :  Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  •  :1405-41-0
  •  :C₂₁H₄₃N₅O₇.H₂SO₄
  •  :575.67 g/mol
  • Pack type:
    Bottle
  • Size:
    10 g