Synthware Microliter Sample Syringes, Kemtech America
Syringe, 25 µl, Beveled
Catalog # 76264-148
Supplier: Kemtech America
Supplier #: S371005
Jump to:
Product Details & Documents
These microliter sample syringes are available in two tip types.
- Beveled and blunt tip
- High quality borosilicate glass
- Fixed needle
The anti-coring beveled tip is suitable for easy septum penetration. The blunt tip is suitable for HPLC injection and sample pipetting.
Specifications
- Volume:25 µl
- Description:Syringe
- Tip style:Beveled