Synthware Microliter Sample Syringes, Kemtech America
Syringe, 5 µl, Beveled
76264-144
: S371003
These microliter sample syringes are available in two tip types.
- Beveled and blunt tip
- High quality borosilicate glass
- Fixed needle
The anti-coring beveled tip is suitable for easy septum penetration. The blunt tip is suitable for HPLC injection and sample pipetting.
- Volume:5 µl
- Description:Syringe
- Tip style:Beveled